missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Grade 5V Fluted Fine Particle Filter Paper, 110 mm Circle

Cytiva's Whatman Grade 5V fine particle filter retains particles as small as 2.5 μm for qualitative analysis. This cellulose filter paper is fluted for ease-of-use.

Marke:  Cytiva 1205-110

Weitere Details : Netto-Gewicht : 0.11020kg

Artikelnummer. 17897664

  • 160.00 € / 100 Stück
Verfügbar ab: 10-06-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

  • Has highest nominal particle retention efficiency of fine particles in Whatman qualitative cellulose paper range
  • Slow filtration speed (Herzberg speed of 1420 s)
  • Fluted (pre-pleated) to facilitate use with conical filter funnels
  • Fluted filter paper design decreases filtration time and increases loading capacity
Spezifikation

Spezifikation

Folded (Pre-pleated)
Cellulose
5 mL/min
100/Pk.
Cellulose
2.5 μm
5V
110 mm
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote