missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wnt-5a Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 13 publications
Marke: Novus Biologicals NBP2-24752
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Wnt-5a Polyclonal specifically detects Wnt-5a in Human, Mouse, Rat, Bovine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Spezifikation
| Wnt-5a | |
| Polyclonal | |
| Unconjugated | |
| PBS pH 8.5 with 0.05% Sodium Azide | |
| hWNT5A, protein Wnt-5a, wingless-type MMTV integration site family, member 5A, WNT-5A protein | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| The amino acid sequence used as immunogen for the Wnt5A antibody is 100% homologous in human, cow, monkey, rat and mouse, 94% in opossum, 88% in xenopus, 81% in chimpanzee and 76% homologous in pufferfish. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 0.5-2 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 5-10 ug/ml, Immunohistochemistry-Frozen reported by customer review | |
| NP_003383 | |
| WNT5A | |
| A synthetic peptide corresponding to amino acids 190-230 of human Wnt-5a was used as the immunogen for this antibody, GenBank no NP_003383.2. | |
| 0.1 mg | |
| Cancer Stem Cells, Signal Transduction, Stem Cell Signaling Pathway, Wnt Signaling Pathway | |
| 7474 | |
| Human, Mouse, Rat, Bovine, Monkey | |
| Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur