missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wnt-5a Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 13 publications
190.00 € - 550.00 €
Spezifikation
| Antigen | Wnt-5a |
|---|---|
| Konzentration | 1.0 mg/mL |
| Verdünnung | Western Blot 0.5-2 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 5-10 ug/ml, Immunohistochemistry-Frozen reported by customer review |
| Anwendungen | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Klassifikation | Polyclonal |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
18009585
|
Novus Biologicals
NBP2-24752SS |
0.025 mg |
190.00 €
25 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
18748253
|
Novus Biologicals
NBP2-24752 |
0.1 mg |
550.00 €
0.10 Milligramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
Wnt-5a Polyclonal specifically detects Wnt-5a in Human, Mouse, Rat, Bovine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Spezifikation
| Wnt-5a | |
| Western Blot 0.5-2 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 5-10 ug/ml, Immunohistochemistry-Frozen reported by customer review | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Bovine, Monkey | |
| NP_003383 | |
| 7474 | |
| A synthetic peptide corresponding to amino acids 190-230 of human Wnt-5a was used as the immunogen for this antibody, GenBank no NP_003383.2. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 1.0 mg/mL | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer Stem Cells, Signal Transduction, Stem Cell Signaling Pathway, Wnt Signaling Pathway | |
| PBS pH 8.5 with 0.05% Sodium Azide | |
| hWNT5A, protein Wnt-5a, wingless-type MMTV integration site family, member 5A, WNT-5A protein | |
| WNT5A | |
| IgG | |
| Affinity purified | |
| The amino acid sequence used as immunogen for the Wnt5A antibody is 100% homologous in human, cow, monkey, rat and mouse, 94% in opossum, 88% in xenopus, 81% in chimpanzee and 76% homologous in pufferfish. |
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts