Learn More
Xanthanhydrid, Thermo Scientific Chemicals
Catalog Number p-7214811
Click to view available options
Menge:
5 g
25 g
6846-35-1
C2H2N2S3
150.23
YWZHEXZIISFIDA-UHFFFAOYSA-N
5-Amino-3H-1,2,4-dithiazol-3-thion
NC1=NC(=S)SS1
6846-35-1
C2H2N2S3
150.23
YWZHEXZIISFIDA-UHFFFAOYSA-N
5-Amino-3H-1,2,4-dithiazol-3-thion
NC1=NC(=S)SS1
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Xanthane hydride
Schmelzpunkt 145.0°C to 150.0°C
Menge 5 g
Formelmasse 150.22