Learn More
YARS Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA553883
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: GNVENNGVLS FIKHVLFPLK SEFVILRDEK WGGNKTYTAY VDLEKDFAAE VVHPGDLKNS VEVALNKLLD PIREKFNTPA LKKLASAAYP DPSKQKPMAK GPAKNSEPEE VIPSRLDIRV GKIITVEKHP DAD Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 93%.
Aminoacyl-tRNA synthetases catalyze the aminoacylation of tRNA by their cognate amino acid. Because of their central role in linking amino acids with nucleotide triplets contained in tRNAs, aminoacyl-tRNA synthetases are thought to be among the first proteins that appeared in evolution. Tyrosyl-tRNA synthetase belongs to the class I tRNA synthetase family. Cytokine activities have also been observed for the human tyrosyl-tRNA synthetase, after it is split into two parts, an N-terminal fragment that harbors the catalytic site and a C-terminal fragment found only in the mammalian enzyme. The N-terminal fragment is an interleukin-8-like cytokine, whereas the released C-terminal fragment is an EMAP II-like cytokine.Spezifikation
YARS | |
Polyclonal | |
Unconjugated | |
YARS1 | |
AL024047; AU018965; CMTDIC; tyrosine tRNA ligase 1, cytoplasmic; tyrosine--tRNA ligase, cytoplasmic; Tyrosine--tRNA ligase, cytoplasmic, N-terminally processed; tyrosyl trna synthetase; tyrosyl--tRNA ligase; tyrosyl-tRNA synthetase; tyrosyl-tRNA synthetase, cytoplasmic; tyrRS; Yars; YRS; YTS | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
107271, 313047, 8565 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P54577, Q4KM49, Q91WQ3 | |
YARS1 | |
Recombinant protein corresponding to Human YARS | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Research Use Only