missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YIF1A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA553240
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: ALMYFIVRSL RTAALGPDSM GGPVPRQRLQ Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 87%.
YIF1A has a possible role in transport between endoplasmic reticulum and Golgi.Spezifikation
YIF1A | |
Polyclonal | |
Unconjugated | |
YIF1A | |
5330422J23Rik; 54TM; 54TMp; FinGER7; HYIF1P; liver protein; protein YIF1A; putative Rab5-interacting protein; putative transmembrane protein 54TMp; putative transmembrane protein; homolog of yeast Golgi membrane protein Yif1p (Yip1p-interacting factor); Yif1; YIF1A; Yif1p; Yip1 interacting factor homolog A; Yip1 interacting factor homolog A (S. cerevisiae); Yip1 interacting factor homolog A, membrane trafficking protein; YIP1-interacting factor homolog A; Yip1p-interacting factor | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10897 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O95070 | |
YIF1A | |
Recombinant protein corresponding to Human YIF1A | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only