Learn More
YIPF1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA543347
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: HLGEKTYHYV PEFRKVSIAA TIIYAYAWLV PLALWGFLMW RNSKVMNIVS Sequence homology: Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 100%
YIPF1 Polyclonal antibody specifically detects YIPF1 in Human samples. It is validated for Western Blot.Spezifikation
YIPF1 | |
Polyclonal | |
Unconjugated | |
YIPF1 | |
C030002N13Rik; DJ167A19.1; FinGER1; protein YIPF1; Yip1 domain family member 1; Yip1 domain family, member 1; YIP1 family member 1; Yipf1; YIPF1-like protein | |
Rabbit | |
Affinity Chromatography | |
RUO | |
54432 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9Y548 | |
YIPF1 | |
Synthetic peptide directed towards the middle region of human YIPF1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |