Learn More
YMC Group YMC-Actus Triart Prep C18-S, semi-preparative HPLC-column (20.0mmID), 12 nm, S-10 µm, 250x20.0mm
Marke:  YMC Group TAS12S11-2520WX
Encompass
YMC-Actus Triart Prep C18-S, semi-preparative HPLC-column (20.0mmID), 12 nm, S-10 µm, 250x20.0mm
Artikelnummer. 15164138
  • 4860.00 € / 1 Stück