Learn More
Ymer Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA531211
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: 293T, A431, HeLa. Predicted reactivity: Mouse (88%), Rat (89%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This protein is a bifunctional mRNA-capping enzyme exhibiting RNA 5'-triphosphatase activity in the N-terminal part and mRNA guanylyltransferase activity in the C-terminal part. Catalyzes the first two steps of cap formation: by removing the gamma-phosphate from the 5'-triphosphate end of nascent mRNA to yield a diphosphate end, and by transferring the gmp moiety of GTP to the 5'-diphosphate terminus.Spezifikation
Ymer | |
Polyclonal | |
Unconjugated | |
Ccdc50 | |
C3orf6; C3orf6h; Ccdc50; coiled-coil domain containing 50; coiled-coil domain-containing protein 50; DFNA44; Protein Ymer; putative C3orf6 protein homolog; YMER; Ymer protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
152137 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q8IVM0 | |
Ccdc50 | |
Recombinant fragment corresponding to a region within amino acids 1 and 187 of Human Ymer | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |