Learn More
YOD1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA555851
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: QRNFPDPDTP PLTIFSSNDD IVLVQALELA DEARRRRQFT DVNRFTLRCM VCQKGLTGQA EAREHAKETG HTNFGEV Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.
Deubiquitinating enzymes (DUBs) are proteases that specifically cleave ubiquitin linkages, negating the action of ubiquitin ligases. DUBA8 belongs to a DUB subfamily characterized by an ovarian tumor (OTU) domain.Spezifikation
YOD1 | |
Polyclonal | |
Unconjugated | |
YOD1 | |
9930028C20Rik; DUBA8; DUBA-8; fe01a10; HIN7; HIN-7; HIV-1-induced protease 7; HIV-induced protein-7-like protease; hsHIN7; hypothetical protein LOC550411; OTU domain containing 2; OTU domain-containing protein 2; OTUD2; PRO0907; Ubiquitin thioesterase OTU1; wu:fe01a10; Yod1; YOD1 deubiquitinase; YOD1 OTU deubiquinating enzyme 1 homolog; YOD1 OTU deubiquinating enzyme 1 homolog ( yeast); YOD1 OTU deubiquitinating enzyme 1 homologue; YOD1 OTU deubiquitinating enzyme 1 homologue (S. cerevisiae); zgc:112182 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55432 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5VVQ6 | |
YOD1 | |
Recombinant protein corresponding to Human YOD1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only