Learn More
YTHDC1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA557670
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: LVSKPLSSSV SNNKRIVSTK GKSATEYKNE EYQRSERNKR LDADRKIRLS SSASREPYKN QPEKTCVRKR DPERRAKSPT PDGSERIGLE VD Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 92%.
May be part of a signal transduction pathway that influences splice site selection. Target protein binds nuclear transcriptosomal complex scaffold attachment factor B (SAF-B) and Sam68; may play a role in splice site selection and alternative splicing.Spezifikation
YTHDC1 | |
Polyclonal | |
Unconjugated | |
YTHDC1 | |
A730098D12Rik; C80342; KIAA1966; mKIAA1966; putative splicing factor YT521; RA301-binding protein; Splicing factor YT521; splicing factor YT521-B; YT521; YT521-B; YTH domain containing 1; YTH domain-containing protein 1; YTHDC1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
91746 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.14 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96MU7 | |
YTHDC1 | |
Recombinant protein corresponding to Human YTHDC1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only