missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YTHDF2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA563756
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: PSQALVQNIG QPTQGSPQPV GQQANNSPPV AQASVGQQTQ Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 95%.
Specifically recognizes and binds N6-methyladenosine (m6A)-containing RNAs, and regulates mRNA stability. M6A is a modification present at internal sites of mRNAs and some non-coding RNAs and plays a role in the efficiency of mRNA splicing, processing and stability. Acts as a regulator of mRNA stability: binding to m6A-containing mRNAs results in the localization to mRNA decay sites, such as processing bodies (P-bodies), leading to mRNA degradation. Also acts as a promoter of cap-independent mRNA translation following heat shock stress: upon stress, relocalizes to the nucleus and specifically binds mRNAs with some m6A methylation mark at their 5'-UTR, protecting demethylation of mRNAs by FTO, thereby promoting cap-independent mRNA translation.Spezifikation
YTHDF2 | |
Polyclonal | |
Unconjugated | |
YTHDF2 | |
9430020E02Rik; CAHL; CLL-associated antigen KW-14; HGRG8; high glucose-regulated protein 8; high-glucose-regulated protein 8; LOW QUALITY PROTEIN: YTH domain-containing family protein 2; NY-REN-2; renal carcinoma antigen NY-REN-2; YTH domain family 2; YTH domain family protein 2; YTH domain family, member 2; YTH domain-containing family protein 2; YTH N(6)-methyladenosine RNA binding protein 2; YTH N6-methyladenosine RNA binding protein 2; yth2; YTHDF2; zgc:56224 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51441 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y5A9 | |
YTHDF2 | |
Recombinant protein corresponding to Human YTHDF2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only