Learn More
Eurofins DiscoverX ZAP-70 Protein, Active
Beschreibung
ZA70 (Tyrosine Protein kinase ZAP-70, 70 kDa zeta-associated protein, Syk-related tyrosine kinase) is a cytosolic protein tyrosine kinase (PTK) and a member of the Syk family of proteins. ZA70 is expressed in human and murine thymocyte subsets, B cells, and peripheral T cell subsets and associates with zeta-chain to undergo tyrosine phosphorylation following TCR stimulation. ZA70 inhibitors may be useful as immunosuppressants.
GenBank L05148
Spezifikation
Spezifikation
| Zusammensetzung | mg/mL of enzyme in of 25 mMHepes/NaOH pH 7.4, 150 mM NaCl, 0.5 mM DTT, 50% glycerol. Liquid at -20°C. |
| Molekulargewicht | 70.8 kDa |
| Menge | 20 μg |
| Spezies | Human |
| Rekombinant | Recombinant |
| Proteinmarkierung | C-terminal 6His |
| Expressionssystem | Recombinant enzyme expressed in Sf21 insect cells |
| Proteinform | Full-length |
| Reinheits- oder Qualitätsgrad | ≥66% |
| Protein | ZAP-70, active |