Learn More
ZBTB20 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA553496
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: SVEQQFGPGA ARDSQAEPTQ PEQAAEAPAE GGPQTNQLET GASSPERSNE VEMDSTVITV SNSSDKSVLQ QPSVNTSIGQ PLPSTQLYLR QTETLTSNLR MPLTLTSNTQ VIGTAGNTYL PALFTTQPAG SGPKPFLFSL PQPLAGQQTQ Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 90%.
This gene, which was initially designated as dendritic cell-derived BTB/POZ zinc finger (DPZF), belongs to a family of transcription factors with an N-terminal BTB/POZ domain and a C-terminal DNA-bindng zinc finger domain. The BTB/POZ domain is a hydrophobic region of approximately 120 aa which mediates association with other BTB/POZ domain-containing proteins. This gene acts as a transcriptional repressor and plays a role in many processes including neurogenesis, glucose homeostasis, and postnatal growth. Mutations in this gene have been associated with Primrose syndrome as well as the 3q13.31 microdeletion syndrome. Alternative splicing results in multiple transcript variants encoding distinct isoforms. [provided by RefSeq, Feb 2017]Spezifikation
ZBTB20 | |
Polyclonal | |
Unconjugated | |
ZBTB20 | |
1300017A20Rik; 7330412A13Rik; A930017C21Rik; BTB/POZ domain zinc finger factor HOF; BTB/POZ zinc finger protein DPZF; D16Wsu73e; dendritic cell-derived BTB/POZ zinc finger; Dendritic-derived BTB/POZ zinc finger protein; DPZF; HOF; Oda8; ODA-8S; POZ/zinc finger transcription factor ODA-8; PRIMS; RGD1560387; Zbtb20; Zfp288; zinc finger and BTB domain containing 20; zinc finger and BTB domain-containing protein 20; Zinc finger protein 288; ZNF288 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26137 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9HC78 | |
ZBTB20 | |
Recombinant protein corresponding to Human ZBTB20 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only