missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF251 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 25601-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
ZNF251 may be involved in transcriptional regulation.Spezifikation
ZNF251 | |
Polyclonal | |
Unconjugated | |
ZNF251 | |
zinc finger protein 251, ZNF251 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
71591, 90987 | |
-20°C | |
Liquid |
Immunofluorescence, Western Blot, Immunocytochemistry | |
0.43 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9BRH9 | |
Zfp251, ZNF251 | |
ZNF251 Fusion Protein Ag22347 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |