missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF606 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 26128-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
May act as a transcriptional repressor.Spezifikation
ZNF606 | |
Polyclonal | |
Unconjugated | |
ZNF606 | |
KIAA1852, Zinc finger protein 328, zinc finger protein 606, ZNF328, ZNF606 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
67370, 80095 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation | |
0.14 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8WXB4 | |
Zfp606, ZNF606 | |
ZNF606 Fusion Protein Ag23589 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |