CAS RN 1000669-72-6
CAS RN 1000669-72-6
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
1
von
1
Ergebnisse
1
KW-2449, MedChemExpress
MedChemExpress KW-2449 is a multi-targeted kinase inhibitor of FLT3, ABL, ABLT315I and Aurora kinase with IC50s of 6.6, 14, 4 and 48 nM, respectively.