CAS RN 1005883-72-6
CAS RN 1005883-72-6
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
1
von
1
Ergebnisse
1
Z433927330, MedChemExpress
MedChemExpress Z433927330 is a potent and selective inhibitor of Aquaporin-7 (AQP7), less potently inhibits AQP3 and AQPs9, with IC50s of ∼0.2 μM, ∼0.7 μM and ∼1.1 μM for mAQP7, mAQP3 and mAQP9, respectively.