CAS RN 1010073-75-2
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
1
von
1
Ergebnisse
1
CCX354, MedChemExpress
MedChemExpress CCX354 is an antagonist of CCR1, with anti-inflammatory activity.