CAS RN 1019331-10-2
CAS RN 1019331-10-2
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
1
von
1
Ergebnisse
1
S0859, MedChemExpress
MedChemExpress S0859 is a selective, high-affinity generic Na+/HCO3- transporter (NBC) inhibitor. S0859 reversibly inhibits NBC-mediated intracellular pH (pHi) recovery (Ki=1.7 μM, full inhibition at approximately 30 μM).