CAS RN 1033040-23-1
CAS RN 1033040-23-1
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
1
von
1
Ergebnisse
1
TPO agonist 1, MedChemExpress
MedChemExpress TPO agonist 1 is a thrombopoietin (TPO) agonist extracted from patent WO2008134338A1, compound TPO mimetic. It would be useful as promoters of thrombopoiesis and megakaryocytopoiesis to treat thrombocytopenia.