CAS RN 1037792-44-1
CAS RN 1037792-44-1
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
1
von
1
Ergebnisse
1
MBX-2982, MedChemExpress
MedChemExpress MBX-2982 is a selective, orally-available G protein-coupled receptor 119 (GPR119) agonist.