CAS RN 1227163-84-9
CAS RN 1227163-84-9
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
1
von
1
Ergebnisse
1
AZD3839 free base, MedChemExpress
MedChemExpress AZD3839 free base is a potent and selective orally active, brain-permeable BACE1 inhibitor (Ki=26 nM).