CAS RN 128312-11-8
IUPAC Name:
[3-(methylsulfanyl)phenyl]boronic acid
Synonyme:
3-(methylsulfanyl)phenylboronic acid
Molekulargewicht (g/mol):
168.02
Summenformel:
C7H9BO2S
InChi Key:
TYVPOLHSKGEXIH-UHFFFAOYSA-N
SMILES:
CSC1=CC=CC(=C1)B(O)O
1
–
1
von
1
Ergebnisse
1
3-(Methylthio)benzenboronsäure, 97 %, Thermo Scientific™
CAS: 128312-11-8 Summenformel: C7H9BO2S Molekulargewicht (g/mol): 168.017 MDL-Nummer: MFCD01319105 InChI-Schlüssel: TYVPOLHSKGEXIH-UHFFFAOYSA-N Synonym: 3-methylthio phenylboronic acid,3-methylthio phenyl boronic acid,3-meththio phenylboronic acid,3-methylthiophenylboronic acid,3-methylthio benzeneboronic acid,3-methylsulfanyl phenylboronic acid,3-boronothioanisole,3-methylthiobenzeneboronic acid,4-methylthiophenyl boronic acid,3-methylthio phenyl-boronic acid PubChem CID: 2734383 IUPAC-Name: (3-methylsulfanylphenyl)Boronsäure SMILES: B(C1=CC(=CC=C1)SC)(O)O