CAS RN 1317-38-0
CAS RN 1317-38-0
Kupfer(II)-oxid, 97 %, Thermo Scientific Chemicals
CAS: 1317-38-0 Summenformel: CuO Molekulargewicht (g/mol): 79.55 MDL-Nummer: MFCD00010979 InChI-Schlüssel: KKCXRELNMOYFLS-UHFFFAOYSA-N Synonym: cupric oxide,copper ii oxide,copper oxide,copper monoxide,banacobru ol,chrome brown,copper brown,copper monooxide,black copper oxide PubChem CID: 14829 SMILES: [O--].[Cu++]
Kupfer(II)-oxid, 99+ %, Thermo Scientific Chemicals
CAS: 1317-38-0 Summenformel: CuO Molekulargewicht (g/mol): 79.55 MDL-Nummer: MFCD00010979 InChI-Schlüssel: KKCXRELNMOYFLS-UHFFFAOYSA-N Synonym: cupric oxide,copper ii oxide,copper oxide,copper monoxide,banacobru ol,chrome brown,copper brown,copper monooxide,black copper oxide PubChem CID: 14829 SMILES: [O--].[Cu++]
Kupfer(II)-oxid, 99.995 % (Metallbasis), Thermo Scientific Chemicals, Puratronic™
CAS: 1317-38-0 Summenformel: CuO Molekulargewicht (g/mol): 79.55 MDL-Nummer: MFCD00010979 InChI-Schlüssel: KKCXRELNMOYFLS-UHFFFAOYSA-N Synonym: cupric oxide,copper ii oxide,copper oxide,copper monoxide,banacobru ol,chrome brown,copper brown,copper monooxide,black copper oxide PubChem CID: 14829 IUPAC-Name: Oxokupfer SMILES: [O--].[Cu++]
Kupfer(II)-oxid, Nanopulver, Thermo Scientific Chemicals
CAS: 1317-38-0 Summenformel: CuO Molekulargewicht (g/mol): 79.55 MDL-Nummer: MFCD00010979 InChI-Schlüssel: KKCXRELNMOYFLS-UHFFFAOYSA-N Synonym: cupric oxide,copper ii oxide,copper oxide,copper monoxide,banacobru ol,chrome brown,copper brown,copper monooxide,black copper oxide PubChem CID: 14829 SMILES: [O--].[Cu++]
Kupfer(II)-oxid, 99.7 % (Metallbasis), Thermo Scientific Chemicals
CAS: 1317-38-0 Summenformel: CuO Molekulargewicht (g/mol): 79.55 MDL-Nummer: MFCD00010979 InChI-Schlüssel: KKCXRELNMOYFLS-UHFFFAOYSA-N Synonym: cupric oxide,copper ii oxide,copper oxide,copper monoxide,banacobru ol,chrome brown,copper brown,copper monooxide,black copper oxide PubChem CID: 14829 IUPAC-Name: Oxokupfer SMILES: [O--].[Cu++]
Kupfer(II)-oxid, 99.9995 % (Metallbasis), Thermo Scientific Chemicals
CAS: 1317-38-0 Summenformel: CuO Molekulargewicht (g/mol): 79.55 MDL-Nummer: MFCD00010979 InChI-Schlüssel: KKCXRELNMOYFLS-UHFFFAOYSA-N Synonym: cupric oxide,copper ii oxide,copper oxide,copper monoxide,banacobru ol,chrome brown,copper brown,copper monooxide,black copper oxide PubChem CID: 14829 IUPAC-Name: Oxokupfer SMILES: [O--].[Cu++]
Kupfer(II)-oxid, ACS, 99.0 % min., Thermo Scientific Chemicals
CAS: 1317-38-0 Summenformel: CuO Molekulargewicht (g/mol): 79.55 MDL-Nummer: MFCD00010979 InChI-Schlüssel: KKCXRELNMOYFLS-UHFFFAOYSA-N Synonym: cupric oxide,copper ii oxide,copper oxide,copper monoxide,banacobru ol,chrome brown,copper brown,copper monooxide,black copper oxide PubChem CID: 14829 SMILES: [O--].[Cu++]
Kupfer(II)-oxid, ACS-Reagenz, Pulver, Thermo Scientific Chemicals
CAS: 1317-38-0 Summenformel: CuO Molekulargewicht (g/mol): 79.55 MDL-Nummer: MFCD00010979 InChI-Schlüssel: KKCXRELNMOYFLS-UHFFFAOYSA-N Synonym: cupric oxide,copper ii oxide,copper oxide,copper monoxide,banacobru ol,chrome brown,copper brown,copper monooxide,black copper oxide PubChem CID: 14829 SMILES: [O--].[Cu++]
Kupfer(II)-oxid, 97 %, Thermo Scientific Chemicals
CAS: 1317-38-0 Summenformel: CuO Molekulargewicht (g/mol): 79.55 MDL-Nummer: MFCD00010979 InChI-Schlüssel: KKCXRELNMOYFLS-UHFFFAOYSA-N Synonym: cupric oxide,copper ii oxide,copper oxide,copper monoxide,banacobru ol,chrome brown,copper brown,copper monooxide,black copper oxide PubChem CID: 14829 IUPAC-Name: Oxokupfer SMILES: [O--].[Cu++]
Kupfer(II)-oxid, ACS, Thermo Scientific Chemicals
CAS: 1317-38-0 Summenformel: CuO Molekulargewicht (g/mol): 79.55 MDL-Nummer: MFCD00010979 InChI-Schlüssel: KKCXRELNMOYFLS-UHFFFAOYSA-N Synonym: cupric oxide,copper ii oxide,copper oxide,copper monoxide,banacobru ol,chrome brown,copper brown,copper monooxide,black copper oxide PubChem CID: 14829 SMILES: [O--].[Cu++]
Kupfer(II)-oxid, technisch, Pulver, Fisher Chemical
CAS: 1317-38-0 Summenformel: CuO Molekulargewicht (g/mol): 79.55 MDL-Nummer: MFCD00010979 InChI-Schlüssel: KKCXRELNMOYFLS-UHFFFAOYSA-N IUPAC-Name: Oxokupfer SMILES: [O--].[Cu++]
Kupfer(II)-oxid, 98 %, Honeywell™ Fluka™
CAS: 1317-38-0 Summenformel: CuO Molekulargewicht (g/mol): 79.55 MDL-Nummer: MFCD00010979 InChI-Schlüssel: KKCXRELNMOYFLS-UHFFFAOYSA-N Synonym: Kupferoxid,Kupfer(II)-oxid,Kupfermonoxid,Chrombraun,Kupferbraun,Kupfermonooxid,Schwarzes Kupferoxid,copper monooxide,black copper oxide PubChem CID: 14829 SMILES: [O--].[Cu++]
Kupfer(II)-oxid, ≥99 % (RT), Granular, Honeywell™ Fluka™
CAS: 1317-38-0 Summenformel: CuO Molekulargewicht (g/mol): 79.55 MDL-Nummer: MFCD00010979 InChI-Schlüssel: KKCXRELNMOYFLS-UHFFFAOYSA-N Synonym: Kupferoxid,Kupfer(II)-oxid,Kupfermonoxid,Chrombraun,Kupferbraun,Kupfermonooxid,Schwarzes Kupferoxid,copper monooxide,black copper oxide PubChem CID: 14829 SMILES: [O--].[Cu++]
Copper(Ii) Oxide, ≥99.0%, ACS Reagent, Honeywell™ Fluka™
CAS: 1317-38-0 Summenformel: CuO Molekulargewicht (g/mol): 79.55 MDL-Nummer: MFCD00010979 InChI-Schlüssel: KKCXRELNMOYFLS-UHFFFAOYSA-N Synonym: cupric oxide,copper ii oxide,copper oxide,copper monoxide,banacobru ol,chrome brown,copper brown,copper monooxide,black copper oxide PubChem CID: 14829 IUPAC-Name: copper(2+) oxidandiide SMILES: [O--].[Cu++]
- 1
- 2