CAS RN 1332295-35-8
CAS RN 1332295-35-8
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
1
von
1
Ergebnisse
1
Nav1.7-IN-2, MedChemExpress
MedChemExpress Nav1.7-IN-2 is an inhibitor of voltage-gated sodium channels (Nav), in particular Nav 1.7, with IC50 of 80 nM.