CAS RN 1357389-11-7
CAS RN 1357389-11-7
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
2
von
2
Ergebnisse
1
Tocris Bioscience™ RGFP 966
Potent and selective HDAC3 inhibitor (IC50 = 80 nM)
2
RGFP966, MedChemExpress
MedChemExpress RGFP966 is a highly selective HDAC3 inhibitor with an IC50 of 80 nM and shows no inhibition to other HDACs at concentrations up to 15 μM. RGFP966 can penetrate the blood brain barrier (BBB).