CAS RN 142063-30-7
IUPAC Name:
sodium 2,4-dimethylbenzene-1-sulfonate hydrate
Synonyme:
sodium 2,4-dimethylbenzenesulfonate hydrate
Molekulargewicht (g/mol):
226.22
Summenformel:
C8H11NaO4S
InChi Key:
LAOKFSDVOMOLJF-UHFFFAOYSA-M
SMILES:
O.[Na+].CC1=CC(C)=C(C=C1)S([O-])(=O)=O