CAS RN 1435934-25-0
CAS RN 1435934-25-0
IUPAC Name:
7-[4-(4-methylpiperazin-1-yl)cyclohexyl]-5-(4-phenoxyphenyl)-7H-pyrrolo[2,3-d]pyrimidin-4-amine trihydrochloride
Synonyme:
7-[4-(4-methylpiperazin-1-yl)cyclohexyl]-5-(4-phenoxyphenyl)pyrrolo[2,3-d]pyrimidin-4-amine trihydrochloride
Molekulargewicht (g/mol):
592.01
Summenformel:
C29H37Cl3N6O
InChi Key:
ALRMEQIQFCUAMR-UHFFFAOYSA-N
SMILES:
Cl.Cl.Cl.CN1CCN(CC1)C1CCC(CC1)N1C=C(C2=C(N)N=CN=C12)C1=CC=C(OC2=CC=CC=C2)C=C1
1
–
2
von
2
Ergebnisse
2
A 419259 trihydrochloride, MedChemExpress
MedChemExpress A 419259 trihydrochloride is a Src family kinases inhibitor with IC50s of 9 nM, 3 nM and 3 nM for Src, Lck and Lyn, respectively.