CAS RN 154012-15-4
IUPAC Name:
ethyl 4,6-dichloro-5-nitropyridine-3-carboxylate
Synonyme:
ethyl 4,6-dichloro-5-nitropyridine-3-carboxylate
Molekulargewicht (g/mol):
265.05
Summenformel:
C8H6Cl2N2O4
InChi Key:
VMNMZVKEDHEOEJ-UHFFFAOYSA-N
SMILES:
CCOC(=O)C1=CN=C(Cl)C(=C1Cl)[N+]([O-])=O