CAS RN 166410-37-3
CAS RN 166410-37-3
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
1
von
1
Ergebnisse
1
Fmoc-1,6-diaminohexane, MedChemExpress
MedChemExpress Fmoc-1,6-diaminohexane is an analog of Osw-1 which has the potential for Alzheimer's disease and cancer treatment from patent US 20140135279 A1.