CAS RN 16774-21-3
CAS RN 16774-21-3
Cer(IV)-ammoniumnitrat, 98+ %, Thermo Scientific Chemicals
CAS: 16774-21-3 Summenformel: H8CeN8O18 MDL-Nummer: MFCD00151121 Synonym: Ammonium cerium(IV) nitrate; CAN
Ammoniumcer(IV)-nitrat, 99 %, Thermo Scientific Chemicals
CAS: 16774-21-3 Summenformel: H8CeN8O18 Molekulargewicht (g/mol): 548.22 MDL-Nummer: MFCD00151121 Synonym: Ceric ammonium nitrate,CAN
Cer(IV) Ammoniumnitrat, ACS, 98.5 % min., Thermo Scientific Chemicals
CAS: 16774-21-3 Summenformel: (NH4)2Ce(NO3)6 MDL-Nummer: MFCD00151121
Ammoniumcerium(IV)-Nitrat, zertifiziertes AR für Analysen, Fisher Chemical
CAS: 16774-21-3 Summenformel: CeH8N8O18 Molekulargewicht (g/mol): 548.22 MDL-Nummer: 151121 InChI-Schlüssel: WIBGOERAEYJBOT-UHFFFAOYSA-N IUPAC-Name: λ⁴-Cerium(4+) bis(Salpetersäure)-Diamintetranitrat SMILES: N.N.[Ce+4].O[N+]([O-])=O.O[N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Ammoniumcer(IV)-nitrat, Thermo Scientific Chemicals
CAS: 16774-21-3 Summenformel: H8CeN8O18 Molekulargewicht (g/mol): 548.22 Synonym: Ceric ammonium nitrate,CAN
Cer(IV)-Ammoniumnitrat, 99.5 % (REO), REacton™, Thermo Scientific Chemicals
CAS: 16774-21-3 Summenformel: H8CeN8O18 MDL-Nummer: MFCD00151121
Ammoniumcer(IV)-nitrat, 99 %, ACS-Reagenz, Thermo Scientific Chemicals
CAS: 16774-21-3 Summenformel: H8CeN8O18 Molekulargewicht (g/mol): 548.22 MDL-Nummer: MFCD00151121 Synonym: Ceric ammonium nitrate,CAN
Ammoniumcer-(IV)-Nitratlösung, Honeywell ™
Ammonium Cerium(IV) Nitrate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
Ammonium Cerium(IV) Nitrate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
Ammonium Cerium(IV) Nitrate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
Ammoniumcerium(IV)-nitrat, reinst, SLR, Fisher Chemical
CAS: 16774-21-3 Summenformel: CeH8N8O18 Molekulargewicht (g/mol): 548.22 MDL-Nummer: 151121 InChI-Schlüssel: WIBGOERAEYJBOT-UHFFFAOYSA-N IUPAC-Name: λ⁴-Cerium(4+) bis(Salpetersäure)-Diamintetranitrat SMILES: N.N.[Ce+4].O[N+]([O-])=O.O[N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Ammoniumcer(IV)-nitrat, ACS-Reagenz, ≥98,5 %, Honeywell Fluka™
CAS: 16774-21-3 MDL-Nummer: MFCD00151121
Ammoniumcer(IV)-nitrat, ≥98.0 % (T), purum p.a., Honeywell™
CAS: 16774-21-3 MDL-Nummer: MFCD00151121
- 1
- 2