CAS RN 1698024-73-5
CAS RN 1698024-73-5
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
1
von
1
Ergebnisse
1
ARS-1323, MedChemExpress
MedChemExpress ARS-1323, the racemate of ARS-1620, is a novel inhibitor of mutant K-ras G12C extracted from patent WO 2015054572 A1.