CAS RN 1738-82-5
IUPAC Name:
4-methylbenzene-1-sulfonic acid; benzyl 2-(2-aminoacetamido)acetate
Synonyme:
benzyl 2-(2-aminoacetamido)acetate; para-toluene sulfonate
Molekulargewicht (g/mol):
394.44
Summenformel:
C18H22N2O6S
InChi Key:
RWBJLSORGVEMHA-UHFFFAOYSA-N
SMILES:
CC1=CC=C(C=C1)S(O)(=O)=O.NCC(=O)NCC(=O)OCC1=CC=CC=C1