CAS RN 174722-31-7
CAS RN 174722-31-7
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
3
von
3
Ergebnisse
1
MedChemExpress Rituximab
MedChemExpress Rituximab is an anti-CD20 chimeric monoclonal antibody used to treat certain autoimmune diseases and types of cancer.
2
MedChemExpress Rituximab
MedChemExpress Rituximab is an anti-CD20 chimeric monoclonal antibody used to treat certain autoimmune diseases and types of cancer.
3
MedChemExpress Rituximab
MedChemExpress Rituximab is an anti-CD20 chimeric monoclonal antibody used to treat certain autoimmune diseases and types of cancer.