CAS RN 1784751-20-7
CAS RN 1784751-20-7
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
1
von
1
Ergebnisse
1
CK1-IN-1, MedChemExpress
MedChemExpress CK1-IN-1 is a casein kinase 1 (CK1) inhibitor extracted from patent WO2015119579A1, compound 1c, has IC50s of 15 nM, 16 nM, 73 nM for CK1δ, and CK1ε, p38σ MAPK, respectively.