CAS RN 2026-42-8
CAS RN 2026-42-8
IUPAC Name:
diethyl [(thiophen-2-yl)methyl]phosphonate
Synonyme:
diethyl thiophen-2-ylmethylphosphonate
Molekulargewicht (g/mol):
234.25
Summenformel:
C9H15O3PS
InChi Key:
GOJBUVQDOFHBLF-UHFFFAOYSA-N
SMILES:
CCOP(=O)(CC1=CC=CS1)OCC
1
–
1
von
1
Ergebnisse
1
Diethyl2-Thienylmethylphosphonat, 95%, Thermo Scientific Chemicals
CAS: 2026-42-8 Summenformel: C9H15O3PS Molekulargewicht (g/mol): 234.25 MDL-Nummer: MFCD00128457 InChI-Schlüssel: GOJBUVQDOFHBLF-UHFFFAOYSA-N Synonym: diethyl thiophen-2-ylmethyl phosphonate,diethyl 2-thienylmethyl phosphonate,thiophen-2-ylmethyl-phosphonic acid diethyl ester,diethyl 2-thienylmethylphosphonate,2-diethoxyphosphorylmethyl thiophene,phosphonic acid, 2-thienylmethyl-, diethyl ester,diethyl thiophen-2-ylmethylphosphonate,ksc207o5j,2-diethoxyphosphinylmethyl thiophene PubChem CID: 4185897 IUPAC-Name: 2-(Diethoxyphosphorylmethyl)thiophen SMILES: CCOP(=O)(CC1=CC=CS1)OCC