CAS RN 2140-73-0
CAS RN 2140-73-0
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
2
von
2
Ergebnisse
1
1-Methyl-Inosine, TRC
CAS: 2140-73-0 Summenformel: C11H14N4O5 Molekulargewicht (g/mol): 282.25 Synonym: 1-Methylinosine,N1-Methylinosine; IUPAC-Name: 9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1-methylpurin-6-one SMILES: CN1C=NC2=C(C1=O)N=CN2C3C(C(C(O3)CO)O)O
2
1-Methylinosine, MedChemExpress
MedChemExpress 1-Methylinosine is a modified nucleotide found at position 37 in tRNA 3' to the anticodon of eukaryotic tRNA.