CAS RN 2295-58-1
IUPAC Name:
1-(2,4,6-trihydroxyphenyl)propan-1-one
Synonyme:
flopropione 夫洛丙酮 labrodax ecapron argobyl flopropionum phloropropiophenone labrodax supanate labroda cospanon
Molekulargewicht (g/mol):
182.18
Summenformel:
C9H10O4
InChi Key:
PTHLEKANMPKYDB-UHFFFAOYSA-N
SMILES:
CCC(=O)C1=C(O)C=C(O)C=C1O
1
–
1
von
1
Ergebnisse
1
Flopropione, MedChemExpress
MedChemExpress Flopropione is a 5-HT receptor antagonist and also a catechol-o-methyltransferase (COMT) inhibitor. Flopropione also as an antispasmodic agent.