CAS RN 2325-27-1
IUPAC Name:
N,1,1,1-tetraphenyl-λ⁵-phosphanimine
Synonyme:
N,1,1,1-tetraphenyl-λ⁵-phosphanimine
Molekulargewicht (g/mol):
353.41
Summenformel:
C24H20NP
InChi Key:
PTLOPIHJOPWUNN-UHFFFAOYSA-N
SMILES:
C1=CC=C(C=C1)N=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
von
1
Ergebnisse
1
Tetraphenylphosphinimid, 97 %, Thermo Scientific Chemicals
CAS: 2325-27-1 Summenformel: C24H20NP Molekulargewicht (g/mol): 353.41 MDL-Nummer: MFCD00003044 InChI-Schlüssel: PTLOPIHJOPWUNN-UHFFFAOYSA-N Synonym: tetraphenylphosphine imide,n-triphenylphosphoranylidene aniline,phenylimino triphenylphosphorane,phosphine imide, tetraphenyl,benzenamine, n-triphenylphosphoranylidene,tetraphenylphosphinimine,triphenylphosphine phenylimide,benzenamine,n-triphenylphosphoranylidene,n,1,1,1-tetraphenyl-??-phosphanimine,triphenyl phenylimino PubChem CID: 75352 IUPAC-Name: triphenyl(phenylimino)-$l^{5}-phosphan SMILES: C1=CC=C(C=C1)N=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1