CAS RN 23357-45-1
IUPAC Name:
1,2,3,4-tetrahydronaphthalen-1-ol
Synonyme:
tetrahydronaphthalenol
Molekulargewicht (g/mol):
148.21
Summenformel:
C10H12O
InChi Key:
JAAJQSRLGAYGKZ-UHFFFAOYNA-N
SMILES:
OC1CCCC2=CC=CC=C12