CAS RN 2434629-02-2
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
1
von
1
Ergebnisse
1
Tocris Bioscience™ Pomalidomide 4'-alkylC5-azide
Cereblon ligand with alkyl linker and terminal azide for onward chemistry