CAS RN 245443-52-1
CAS RN 245443-52-1
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
3
von
3
Ergebnisse
1
MedChemExpress Protease-Activated Receptor-4
MedChemExpress Protease-Activated Receptor-4 is the agonist of proteinase-activated receptor-4 (PAR4).
2
MedChemExpress Protease-Activated Receptor-4
MedChemExpress Protease-Activated Receptor-4 is the agonist of proteinase-activated receptor-4 (PAR4).
3
MedChemExpress Protease-Activated Receptor-4
MedChemExpress Protease-Activated Receptor-4 is the agonist of proteinase-activated receptor-4 (PAR4).