CAS RN 252979-43-4
CAS RN 252979-43-4
IUPAC Name:
3-[4-(furan-2-yl)-11-propyl-3,5,6,8,10,11-hexaazatricyclo[7.3.0.0²,⁶]dodeca-1(12),2,4,7,9-pentaen-7-yl]-1-(4-methoxyphenyl)urea
Synonyme:
3-[4-(furan-2-yl)-11-propyl-3,5,6,8,10,11-hexaazatricyclo[7.3.0.0²,⁶]dodeca-1(12),2,4,7,9-pentaen-7-yl]-1-(4-methoxyphenyl)urea
Molekulargewicht (g/mol):
432.44
Summenformel:
C21H20N8O3
InChi Key:
CJRNHKSLHHWUAB-UHFFFAOYSA-N
SMILES:
CCCN1C=C2C(=N1)N=C(NC(=O)NC1=CC=C(OC)C=C1)N1N=C(N=C21)C1=CC=CO1
1
–
1
von
1
Ergebnisse
1
MRE3008F20, MedChemExpress
MedChemExpress MRE3008F20 is a highly potent and selective antagonist of adenosine A3 receptor (AA3R), inhibiting agonist-induced cAMP elevation in resting T lymphocytes with an IC50 of 5 nM.