CAS RN 325970-71-6
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
1
von
1
Ergebnisse
1
L67, MedChemExpress
MedChemExpress L67 is a novel, competitive human DNA ligase inhibitor, inhibits DNA ligases I and III with IC50 of 10 μM and 10 μM.