CAS RN 330461-64-8
CAS RN 330461-64-8
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
1
von
1
Ergebnisse
1
AK-1, MedChemExpress
MedChemExpress AK-1 is a potent, specific and cell-permeable SIRT2 inhibitor, with an IC50 of 12.5 μM.