CAS RN 37779-49-0
IUPAC Name:
methyl 3-oxo-4-phenylbutanoate
Synonyme:
methyl 3-oxo-4-phenylbutanoate
Molekulargewicht (g/mol):
192.21
Summenformel:
C11H12O3
InChi Key:
DTMSEOVTDVSPDO-UHFFFAOYSA-N
SMILES:
COC(=O)CC(=O)CC1=CC=CC=C1