CAS RN 3887-02-3
IUPAC Name:
N,2-dimethylprop-2-enamide
Synonyme:
N,2-dimethylprop-2-enamide
Molekulargewicht (g/mol):
99.13
Summenformel:
C5H9NO
InChi Key:
WFKDPJRCBCBQNT-UHFFFAOYSA-N
SMILES:
CNC(=O)C(C)=C