CAS RN 389621-80-1
CAS RN 389621-80-1
IUPAC Name:
[4-(diethylcarbamoyl)phenyl]boronic acid
Synonyme:
4-(diethylcarbamoyl)phenylboronic acid
Molekulargewicht (g/mol):
221.06
Summenformel:
C11H16BNO3
InChi Key:
ZCGVBHIMRVYWOH-UHFFFAOYSA-N
SMILES:
CCN(CC)C(=O)C1=CC=C(C=C1)B(O)O
1
–
1
von
1
Ergebnisse
1
4-(Diethylcarbamoyl)benzolboronsäure, 98%, Thermo Scientific Chemicals
CAS: 389621-80-1 Summenformel: C11H16BNO3 Molekulargewicht (g/mol): 221.06 MDL-Nummer: MFCD03411949 InChI-Schlüssel: ZCGVBHIMRVYWOH-UHFFFAOYSA-N Synonym: 4-n,n-diethylaminocarbonyl phenylboronic acid,4-diethylcarbamoyl phenylboronic acid,4-diethylcarbamoyl phenyl boronic acid,4-diethylcarbamoyl benzeneboronic acid,boronic acid, 4-diethylamino carbonyl phenyl,4-diethylaminocarbonyl phenylboronic acid,acmc-209j2q,amtb302,n,n-diethyl 4-boronobenzamide,4-diethylcarbamoyl benzeneboronic acid PubChem CID: 2773375 IUPAC-Name: [4-(Diethylcarbamoyl)phenyl]boronsäure SMILES: CCN(CC)C(=O)C1=CC=C(C=C1)B(O)O