CAS RN 39016-05-2
CAS RN 39016-05-2
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
2
von
2
Ergebnisse
1
MedChemExpress C-Peptide, dog
MedChemExpress C-Peptide, dog is a component of proinsulin, released from pancreatic beta cells into blood together with insulin.
2
MedChemExpress C-Peptide, dog
MedChemExpress C-Peptide, dog is a component of proinsulin, released from pancreatic beta cells into blood together with insulin.